Läs mer
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic Acid, TRC
Produktkod. 30137949
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137949 2.5 g
30136642 500 mg
30040506 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137949 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2650402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34598-33-9
Molekylformel C11 H5 F17 O2
Molekylvikt (g/mol) 492.13
Synonym 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid, 2H,2H,3H,3H-Perfluoroundecanoic acid, 3-Perfluorooctylpropionic acid, Heptadecafluoroundecanoic acid
IUPAC-namn 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
LEDER OC(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2,2,3,3H-Perfluoroundecanoic acid
Anteckningar om renhetsgrad GC
Kvantitet 2.5 g
InChI formel InChI=1S/C11H5F17O2/c12-4(13,2-1-3(29)30)5(14,15)6(16,17)7(18,19)8(20,21)9(22,23)10(24,25)11(26,27)28/h1-2H2,(H,29,30)
Formel vikt 492.0018
Procent renhet >95%