missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane, 97%, Thermo Scientific Chemicals

Catalog Number p-4228914
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
331958-90-8
C10H19BO3
198.07
MFCD09878900
KEBDNHHRQNZXMA-UHFFFAOYNA-N
4,4,5,5-tetramethyl-2-tetrahydrofuran-3-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-oxolan-3-yl-1,3,2-dioxaborolane, tetrahydrofuran-3-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-3-furanyl-1,3,2-dioxaborolane, tetrahydrofuran-3-ylboronic acid pinacol ester, tetrahydrofuran-3-yl boronic acid pinacol ester, 2-tetrahydrofuran-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydrofuran
11148297
4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CCOC1
This item is not returnable. View return policy
331958-90-8
C10H19BO3
198.07
MFCD09878900
KEBDNHHRQNZXMA-UHFFFAOYNA-N
4,4,5,5-tetramethyl-2-tetrahydrofuran-3-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-oxolan-3-yl-1,3,2-dioxaborolane, tetrahydrofuran-3-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-tetrahydro-3-furanyl-1,3,2-dioxaborolane, tetrahydrofuran-3-ylboronic acid pinacol ester, tetrahydrofuran-3-yl boronic acid pinacol ester, 2-tetrahydrofuran-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl tetrahydrofuran
11148297
4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CCOC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 198.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Show More Show Less
compliance-icons
Product Identifier
  • 4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'