missing translation for 'onlineSavingsMsg'
Läs mer

4,4'-[(5-Nitro-1,3-phenylene)bis(carbonylimino)]bis-1,3-Benzenedisulfonic Acid Tetrasodium Salt, TRC

Produktkod. 30481873
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481873 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30481873 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN503435100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 4,4'-[(5-Nitro-1,3-phenylene)bis(carbonylimino)]bis-1,3-Benzenedisulfonic Acid Tetrasodium Salt
CAS 709029-71-0
Molekylformel C20H11N3Na4O16S4
Kvantitet 100 mg
Synonym 4,4’-[(5-Nitro-1,3-Phenylene)Bis(Carbonylimino)]Bis-1,3-Benzenedisulfonic Acid Tetrasodium Salt
InChI formel InChI=1S/C20H15N3O16S4.4Na/c24-19(21-15-3-1-13(40(28,29)30)8-17(15)42(34,35)36)10-5-11(7-12(6-10)23(26)27)20(25)22-16-4-2-14(41(31,32)33)9-18(16)43(37,38)39;;;;/h1-9H,(H,21,24)(H,22,25)(H,28,29,30)(H,31,32,33)(H,34,35,36)(H,37,38,39);;;;/q;4*+1/p-4
LEDER O=C(C1=CC(N(=O)=O)=CC(C(NC2=CC=C(S(=O)(O[Na])=O)C=C2S(=O)(O[Na])=O)=O)=C1)NC3=CC=C(S(=O)(O[Na])=O)C=C3S(=O)(O[Na])=O
IUPAC-namn sodium 4,4'-((5-nitroisophthaloyl)bis(azanediyl))bis(benzene-1,3-disulfonate)
Molekylvikt (g/mol) 769.53
Formel vikt 768.86
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.