Läs mer
4-(4,6-Bis(4-((2-ethylhexyloxy)carbonyl)phenylamino)-1,3,5-triazin-2-ylamino)benzoic Acid, TRC
Produktkod. 30331404
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30331404 25 mg
1 options
Produktkod. 30331404 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB43384025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(4,6-Bis(4-((2-ethylhexyloxy)carbonyl)phenylamino)-1,3,5-triazin-2-ylamino)benzoic Acid
CAS 1469870-11-8
Molekylformel C40 H50 N6 O6
Kvantitet 25 mg
Synonym Ethylhexyl Triazone Related Compound B
InChI formel InChI=1S/C40H50N6O6/c1-5-9-11-27(7-3)25-51-36(49)30-15-21-33(22-16-30)42-39-44-38(41-32-19-13-29(14-20-32)35(47)48)45-40(46-39)43-34-23-17-31(18-24-34)37(50)52-26-28(8-4)12-10-6-2/h13-24,27-28H,5-12,25-26H2,1-4H3,(H,47,48)(H3,41,42,43,44,45,46)
LEDER CCCCC(CC)COC(=O)c1ccc(Nc2nc(Nc3ccc(cc3)C(=O)O)nc(Nc4ccc(cc4)C(=O)OCC(CC)CCCC)n2)cc1
IUPAC-namn 4-[[4,6-bis[4-(2-ethylhexoxycarbonyl)anilino]-1,3,5-triazin-2-yl]amino]benzoic acid
Molekylvikt (g/mol) 710.8616
Formel vikt 710.3792