Learn More
4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid, TRC
Product Code. 30401151
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30401151 1 g 1g
30393138 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30401151 Supplier Toronto Research Chemicals Supplier No. TRCF0564501G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic Acid
CAS 1017384-80-3
Molecular Formula C16 H17 F N2 O4
Quantity 1 g
InChI Formula InChI=1S/C16H17FN2O4/c17-11-1-2-12-13(9-11)23-18-16(12)10-5-7-19(8-6-10)14(20)3-4-15(21)22/h1-2,9-10H,3-8H2,(H,21,22)
SMILES OC(=O)CCC(=O)N1CCC(CC1)c2noc3cc(F)ccc23
IUPAC Name 4-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic acid
Molecular Weight (g/mol) 320.316
Formula Weight 320.117