Läs mer
4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis[1-methyl-pyridinium Diiodide (>90%), TRC
Produktkod. 30360987
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30360987 500 mg
30353405 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30360987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC176070500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 647859-37-8
Molekylformel C28 H25 N3 . 2 I
Molekylvikt (g/mol) 657.327
Synonym 4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis[1-methyl-pyridinium Iodide (1:2), 3,6-Bis(1-methyl-4-vinylpyridinium iodine) Carbazole
IUPAC-namn 3,6-bis[2-(1-methylpyridin-1-ium-4-yl)ethenyl]-9H-carbazole;diiodide
LEDER [I-].[I-].C[n+]1ccc(C=Cc2ccc3[nH]c4ccc(C=Cc5cc[n+](C)cc5)cc4c3c2)cc1
Specifikationer
Kemiskt namn eller material 4,4'-[9H-Carbazole-3,6-diyldi-(1E)-2,1-ethenediyl]bis-1-methyl-pyridinium Diiodide
Kvantitet 500 mg
InChI formel InChI=1S/C28H24N3.2HI/c1-30-15-11-21(12-16-30)3-5-23-7-9-27-25(19-23)26-20-24(8-10-28(26)29-27)6-4-22-13-17-31(2)18-14-22;;/h3-20H,1-2H3;2*1H/q+1;;/p-1
Formel vikt 657.014