Läs mer
4-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid, TRC
Produktkod. 30325694
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30325694 10 mg
30320620 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30325694 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61916010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(4-Amino-1-oxoisoindolin-2-yl)-4-carbamoyl Butyric Acid
CAS 2197414-57-4
Molekylformel C13 H15 N3 O4
Kvantitet 10 mg
Synonym 2-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)pentanedioic acid, 5-Amino-4-(4-amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-5-oxopentanoic Acid
InChI formel InChI=1S/C13H15N3O4/c14-9-3-1-2-7-8(9)6-16(13(7)20)10(12(15)19)4-5-11(17)18/h1-3,10H,4-6,14H2,(H2,15,19)(H,17,18)
LEDER NC(=O)C(CCC(=O)O)N1Cc2c(N)cccc2C1=O
IUPAC-namn 5-amino-4-(4-amino-1-oxo-isoindolin-2-yl)-5-oxo-pentanoic acid
Molekylvikt (g/mol) 277.28
Formel vikt 277.1063