Läs mer
4,4'-Azoxydiphenetole, TRC
Produktkod. 30346089
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30346089 250 mg
1 options
Produktkod. 30346089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA965148250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4'-Azoxydiphenetole
CAS 4792-83-0
Molekylformel C16H18N2O3
Kvantitet 250 mg
Synonym Phenetole, 4,4'-azoxydi- (6CI,7CI), 4,4'-Azoxyphenetole, 4,4'-Bis(ethoxy)azoxybenzene, 4,4'-Diethoxyazoxybenzene, ADP, Azoxyphenetole, NSC 142006, PAP, p,p'-Azoxyphenetole, p,p'-Diethoxyazoxybenzene, p,p'-Diethyloxyazoxybenzene
InChI formel InChI=1S/C16H18N2O3/c1-3-20-15-9-5-13(6-10-15)17-18(19)14-7-11-16(12-8-14)21-4-2/h5-12H,3-4H2,1-2H3
LEDER CCOc1ccc(cc1)N=[N+]([O-])c2ccc(OCC)cc2
IUPAC-namn (4-ethoxyphenyl)-(4-ethoxyphenyl)imino-oxidoazanium
Molekylvikt (g/mol) 286.33
Formel vikt 286.1317