missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
4,4'-Benzidine D8 (biphenyl D8) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Benzidine ring-d8 |
| CAS | 92890-63-6 |
| Lösningsmedel eller matris | Acetonitrile |
| Molekylformel | C12 2H8 H4 N2 |
| Kvantitet | 1 mL |
| Synonym | 4,4'-Diamino(biphenyl-d8) |
| InChI formel | InChI=1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2/i1D,2D,3D,4D,5D,6D,7D,8D |
| LEDER | [2H]c1c([2H])c(c([2H])c([2H])c1N)c2c([2H])c([2H])c(N)c([2H])c2[2H] |
| IUPAC-namn | 4-(4-amino-2,3,5,6-tetradeuteriophenyl)-2,3,5,6-tetradeuterioaniline |
| Molekylvikt (g/mol) | 192.29 |
| Visa mer |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?