missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Biphenylyl)-4-oxobutyric acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7040711
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
36330-85-5
C16H14O3
254.285
MFCD00056701
ZPAKPRAICRBAOD-UHFFFAOYSA-N
fenbufen, lederfen, cinopal, bufemid, napanol, 4-4-biphenylyl-4-oxobutyric acid, 3-4-phenylbenzoyl propionic acid, 3-4-biphenylylcarbonyl propionic acid, 4-biphenyl-4-yl-4-oxobutanoic acid, gamma-oxo 1,1'-biphenyl-4-butanoic acid
3335
CHEBI:31599
4-oxo-4-(4-phenylphenyl)butanoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CCC(=O)O
This item is not returnable. View return policy
36330-85-5
C16H14O3
254.285
MFCD00056701
ZPAKPRAICRBAOD-UHFFFAOYSA-N
fenbufen, lederfen, cinopal, bufemid, napanol, 4-4-biphenylyl-4-oxobutyric acid, 3-4-phenylbenzoyl propionic acid, 3-4-biphenylylcarbonyl propionic acid, 4-biphenyl-4-yl-4-oxobutanoic acid, gamma-oxo 1,1'-biphenyl-4-butanoic acid
3335
CHEBI:31599
4-oxo-4-(4-phenylphenyl)butanoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 184°C to 187°C
Quantity 1 g
UN Number UN2811
Formula Weight 254.28
Percent Purity 96%
Chemical Name or Material 4-(4-Biphenylyl)-4-oxobutyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'