missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
4,4'-Bis(4-amino-1-naphthylazo)-2,2'-stilbenedisulfonic Acid (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4,4'-Bis(4-amino-1-naphthylazo)-2,2'-stilbenedisulfonic Acid |
| CAS | 5463-64-9 |
| Molekylformel | C34 H26 N6 O6 S2 |
| Kvantitet | 250 mg |
| Synonym | 2,2'-(1,2-Ethenediyl)bis[5-[2-(4-amino-1-naphthalenyl)diazenyl]benzenesulfonic Acid |
| InChI formel | InChI=1S/C34H26N6O6S2/c35-29-15-17-31(27-7-3-1-5-25(27)29)39-37-23-13-11-21(33(19-23)47(41,42)43)9-10-22-12-14-24(20-34(22)48(44,45)46)38-40-32-18-16-30(36)26-6-2-4-8-28(26)32/h1-20H,35-36H2,(H,41,42,43)(H,44,45,46)/b10-9+,39-37+,40-38+ |
| LEDER | Nc1ccc(N=Nc2ccc(\C=C\c3ccc(cc3S(=O)(=O)O)N=Nc4ccc(N)c5ccccc45)c(c2)S(=O)(=O)O)c6ccccc16 |
| IUPAC-namn | 5-[(E)-(4-aminonaphthalen-1-yl)diazenyl]-2-[(E)-2-[4-[(E)-(4-aminonaphthalen-1-yl)diazenyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid |
| Molekylvikt (g/mol) | 678.74 |
| Formel vikt | 678.1355 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?