Läs mer
4,4'-Bis(isocyanatophenyl)oxide, TRC
Produktkod. 18012420
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012420 100 mg
18023220 250 mg
18051910 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18012420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB448770100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4128-73-8
Molekylformel C14H8N2O3
Molekylvikt (g/mol) 252.22
Synonym 1,1'-Oxybis[4-isocyanatobenzene], 4,4'-Diisocyanatodiphenyl Ether, 4,4'-Oxybis(phenyl Isocyanate), 4-Isocyanatophenyl ether, Diphenyl Ether 4,4'-Diisocyanate, Diphenyl Oxide 4,4'-diisocyanate, Oxybis(4-phenyl Isocyanate);Oxydi-p-phenylene Diisocyanate, Oxydi-p-phenylene Isocyanate
IUPAC-namn 1-isocyanato-4-(4-isocyanatophenoxy)benzene
LEDER O=C=Nc1ccc(Oc2ccc(cc2)N=C=O)cc1
Specifikationer
Kemiskt namn eller material 4,4'-Bis(isocyanatophenyl)oxide
Kvantitet 100 mg
InChI formel InChI=1S/C14H8N2O3/c17-9-15-11-1-5-13(6-2-11)19-14-7-3-12(4-8-14)16-10-18/h1-8H
Formel vikt 252.0535