missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Boc-1-piperazinyl)benzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7039309
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1233243-62-3
C16H24N4O3
320.393
MFCD03791212
XNXLELZXYPNZRJ-UHFFFAOYSA-N
tert-butyl 4-4-z-n'-hydroxycarbamimidoyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-n'-hydroxycarbamimidoyl phenyl piperazine-1-carboxylate, butylaminohydroxyiminomethylphenyltetrahydropyrazinecarboxylate
2763840
tert-butyl 4-[4-(N'-hydroxycarbamimidoyl)phenyl]piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=NO)N
This item is not returnable. View return policy
1233243-62-3
C16H24N4O3
320.393
MFCD03791212
XNXLELZXYPNZRJ-UHFFFAOYSA-N
tert-butyl 4-4-z-n'-hydroxycarbamimidoyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-n'-hydroxycarbamimidoyl phenyl piperazine-1-carboxylate, butylaminohydroxyiminomethylphenyltetrahydropyrazinecarboxylate
2763840
tert-butyl 4-[4-(N'-hydroxycarbamimidoyl)phenyl]piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)C(=NO)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 320.39
Percent Purity 97%
Chemical Name or Material 4-(4-Boc-1-piperazinyl)benzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'