Läs mer
4-((4-Carbamoylpiperidin-1-yl)methyl)benzoic Acid, TRC
Produktkod. 30353857
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353857 25 mg
30350313 100 mg
30353856 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30353857 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18334525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 938143-02-3
Molekylformel C14H18N2O3
Molekylvikt (g/mol) 262.3
Synonym Revefenacin Impurity 5
IUPAC-namn 4-[(4-carbamoylpiperidin-1-yl)methyl]benzoic acid
LEDER NC(=O)C1CCN(Cc2ccc(cc2)C(=O)O)CC1
Specifikationer
Kemiskt namn eller material 4-((4-Carbamoylpiperidin-1-yl)methyl)benzoic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C14H18N2O3/c15-13(17)11-5-7-16(8-6-11)9-10-1-3-12(4-2-10)14(18)19/h1-4,11H,5-9H2,(H2,15,17)(H,18,19)
Formel vikt 262.1317