Läs mer
4-[(4-Chlorophenyl)-2-pyridylmethoxy]piperidine p-Nitrobenzoic Acid Salt, TRC
Produktkod. 30361913
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30361913 10 g
1 options
Produktkod. 30361913 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38038010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-[(4-Chlorophenyl)-2-pyridylmethoxy]piperidine p-Nitrobenzoic Acid Salt
CAS 161558-45-8
Molekylformel C17H19ClN2O.C7H5NO4
Kvantitet 10 g
Synonym 4-Nitrobenzoic acid Compd. With 2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine (1:1), 2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine Mono(4-nitrobenzoate)
InChI formel InChI=1S/C17H19ClN2O.C7H5NO4/c18-14-6-4-13(5-7-14)17(16-3-1-2-10-20-16)21-15-8-11-19-12-9-15;9-7(10)5-1-3-6(4-2-5)8(11)12/h1-7,10,15,17,19H,8-9,11-12H2;1-4H,(H,9,10)
LEDER OC(=O)c1ccc(cc1)[N+](=O)[O-].Clc2ccc(cc2)C(OC3CCNCC3)c4ccccn4
IUPAC-namn 2-[(4-chlorophenyl)-piperidin-4-yloxymethyl]pyridine;4-nitrobenzoic acid
Molekylvikt (g/mol) 469.92
Formel vikt 469.1404