Läs mer
4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, TRC
Produktkod. 30367356
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367356 100 mg
30364358 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367356 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC377860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36159-73-6
Molekylformel C16H13ClO
Molekylvikt (g/mol) 256.73
Synonym 1(2H)-Naphthalenone, 4-(4-chlorophenyl)-3,4-dihydro-, 4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
IUPAC-namn 4-(4-chlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
LEDER Clc1ccc(cc1)C2CCC(=O)c3ccccc23
Specifikationer
Kemiskt namn eller material 4-(4-Chlorophenyl)-3,4-dihydro-1(2H)-naphthalenone
Kvantitet 100 mg
InChI formel InChI=1S/C16H13ClO/c17-12-7-5-11(6-8-12)13-9-10-16(18)15-4-2-1-3-14(13)15/h1-8,13H,9-10H2
Formel vikt 256.0655