Läs mer
4-(4-Chlorophenyl)phenylacetic acid, TRC
Produktkod. 30367034
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367034 250 mg
30366611 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367034 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC428210250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5525-72-4
Molekylformel C14H11ClO2
Molekylvikt (g/mol) 246.69
IUPAC-namn 2-[4-(4-chlorophenyl)phenyl]acetic acid
LEDER OC(=O)Cc1ccc(cc1)c2ccc(Cl)cc2
Specifikationer
Kemiskt namn eller material 4-(4-Chlorophenyl)phenylacetic acid
Kvantitet 250 mg
InChI formel InChI=1S/C14H11ClO2/c15-13-7-5-12(6-8-13)11-3-1-10(2-4-11)9-14(16)17/h1-8H,9H2,(H,16,17)
Formel vikt 246.0448