Läs mer
4-(4-Cyanobenzyl)-1,2,4-triazole, TRC
Produktkod. 30356461
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356461 10 g
30355398 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC96233510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112809-27-5
Molekylformel C10 H8 N4
Molekylvikt (g/mol) 184.2
Synonym 1-(4-Cyanobenzyl)-1,3,4-triazole, 4-(4H-1,2,4-triazol-4-ylmethyl)benzonitrile
IUPAC-namn 4-(1,2,4-triazol-4-ylmethyl)benzonitrile
LEDER N#Cc1ccc(Cn2cnnc2)cc1
Specifikationer
Kemiskt namn eller material 4-(4-Cyanobenzyl)-1,2,4-triazole
Kvantitet 10 g
InChI formel InChI=1S/C10H8N4/c11-5-9-1-3-10(4-2-9)6-14-7-12-13-8-14/h1-4,7-8H,6H2
Formel vikt 184.0749