Läs mer
4,4'-DDE, TRC
Produktkod. 30350986
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350986 500 mg
30351192 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD195205500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72-55-9
Molekylformel C14 H8 Cl4
Molekylvikt (g/mol) 318.03
Synonym Benzene, 1,1'-(dichloroethenylidene)bis[4-chloro- (9CI), Ethylene, 1,1-dichloro-2,2-bis(p-chlorophenyl)- (7CI,8CI), 1,1'-(2,2-Dichloroethenylidene)bis[4-chlorobenzene], 1,1-Bis(4-chlorophenyl)-2,2-dichloroethene, 1,1-Bis(p-chlorophenyl)-2,2-dichloroethylene, 1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene, 1,1-Dichloro-2,2-di(p-chlorophenyl)ethylene, 1,1'-Dichloro-2,2-bis(4-chlorophenyl)ethane, 2,2-Bis(4-chlorophenyl)-1,1-dichloroethene, 2,2-Bis(4-chlorophenyl)-1,1-dichloroethylene, 2,2-Di(p-chlorophenyl)-1,1-dichloroethylene, 2,2-Dichloro-1,1-bis(4-chlorophenyl)ethylene, 4,4'-DDE, 4,4'-Dichlorodiphenyldichloroethylene, DDE, NSC 1153, p,p'-DDE, p,p'-Dichlorodiphenyldichloroethylene
IUPAC-namn 1-chloro-4-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
LEDER ClC(=C(c1ccc(Cl)cc1)c2ccc(Cl)cc2)Cl
Specifikationer
Kemiskt namn eller material 4,4'-DDE
Kvantitet 500 mg
InChI formel InChI=1S/C14H8Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8H
Formel vikt 315.938