Läs mer
4,4'-Diazoaminodibenzenesulfonic Acid Disodium Salt, TRC
Produktkod. 30004737
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004737 1 g
17991585 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30004737 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4171551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56120-28-6
Molekylformel C12 H9 N3 O6 S2 . 2 Na
Molekylvikt (g/mol) 401.33
Synonym Benzenesulfonic acid, 4-[3-(4-sulfophenyl)-1-triazen-1-yl]-, sodium salt (1:2), Benzenesulfonic acid, 4,4'-(1-triazene-1,3-diyl)bis-, disodium salt (9CI), Benzenesulfonic acid, 4,4'-diazoaminodi-, disodium salt (6CI)
IUPAC-namn disodium;4-[(2E)-2-(4-sulfonatophenyl)iminohydrazinyl]benzenesulfonate
LEDER [Na+].[Na+].[O-]S(=O)(=O)c1ccc(NN=Nc2ccc(cc2)S(=O)(=O)[O-])cc1
Specifikationer
Kemiskt namn eller material 4,4'-Diazoaminodibenzenesulfonic Acid Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C12H11N3O6S2.2Na/c16-22(17,18)11-5-1-9(2-6-11)13-15-14-10-3-7-12(8-4-10)23(19,20)21;;/h1-8H,(H,13,14)(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2
Formel vikt 400.9728
Procent renhet >95%