missing translation for 'onlineSavingsMsg'
Learn More

4,4''-Dibromo-p-terphenyl, 98%, Thermo Scientific Chemicals

Catalog Number p-7077521
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
17788-94-2
C18H12Br2
388.10
MFCD00272889
VAIPJQIPFPRJKJ-UHFFFAOYSA-N
4,4-dibromo-p-terphenyl, 4,4-dibromo-1,1':4',1-terphenyl, 1,1':4',1-terphenyl, 4,4-dibromo, acmc-1c6h6, 1,4-bis 4-bromophenyl benzene, 4-bromo-4'-4-bromophenyl-1,1'-biphenyl, 1∼4∼,3∼4∼-dibromo-1∼1∼,2∼1∼:2∼4∼,3∼1∼-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-p-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-1,1 inverted exclamation marka:4 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-terphenyl
12455998
1,4-bis(4-bromophenyl)benzene
BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
This item is not returnable. View return policy
17788-94-2
C18H12Br2
388.10
MFCD00272889
VAIPJQIPFPRJKJ-UHFFFAOYSA-N
4,4-dibromo-p-terphenyl, 4,4-dibromo-1,1':4',1-terphenyl, 1,1':4',1-terphenyl, 4,4-dibromo, acmc-1c6h6, 1,4-bis 4-bromophenyl benzene, 4-bromo-4'-4-bromophenyl-1,1'-biphenyl, 1∼4∼,3∼4∼-dibromo-1∼1∼,2∼1∼:2∼4∼,3∼1∼-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-p-terphenyl, 4,4 inverted exclamation marka inverted exclamation marka-dibromo-1,1 inverted exclamation marka:4 inverted exclamation marka,1 inverted exclamation marka inverted exclamation marka-terphenyl
12455998
1,4-bis(4-bromophenyl)benzene
BrC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 318.0°C to 320.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 388.1
Percent Purity 98%
Chemical Name or Material 4,4''-Dibromo-p-terphenyl
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4''-Dibromo-p-terphenyl, 98%
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'