missing translation for 'onlineSavingsMsg'
Learn More

4',4''-Dibromodiphenylbutadiyne, 97%, Thermo Scientific™

Catalog Number 15204418
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
959-88-6
C16H8Br2
360.05
MFCD00825678
IIERBCCHBFZMQZ-UHFFFAOYSA-N
1,4-bis 4-bromophenyl-1,3-butadiyne, 1,3-butadiyne, 1,4-di 4-bromophenyl, 4,4'-dibromodiphenylbutadiyne, 1,4-bis 4-bromophenyl buta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyl bis 4-bromobenzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene, 1-bromo-4-4-4-bromophenyl buta-1,3-diynyl benzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene #, 1-bromo-4-4-4-bromophenyl buta-1,3-diyn-1-yl benzene
632304
1-bromo-4-[4-(4-bromophenyl)buta-1,3-diynyl]benzene
BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1
This item is not returnable. View return policy
959-88-6
C16H8Br2
360.05
MFCD00825678
IIERBCCHBFZMQZ-UHFFFAOYSA-N
1,4-bis 4-bromophenyl-1,3-butadiyne, 1,3-butadiyne, 1,4-di 4-bromophenyl, 4,4'-dibromodiphenylbutadiyne, 1,4-bis 4-bromophenyl buta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyl bis 4-bromobenzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene, 1-bromo-4-4-4-bromophenyl buta-1,3-diynyl benzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene #, 1-bromo-4-4-4-bromophenyl buta-1,3-diyn-1-yl benzene
632304
1-bromo-4-[4-(4-bromophenyl)buta-1,3-diynyl]benzene
BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1
Melting Point 264.0°C to 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 360.05
Percent Purity 97%
Chemical Name or Material 4',4''-Dibromodiphenylbutadiyne
compliance-icons
Product Identifier
  • 4',4''-Dibromodiphenylbutadiyne, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'