Läs mer
4,4'-Dichlorobenzhydrol, 98%, Thermo Scientific™
Beskrivning
Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .
Applications
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4,4'-Dichlorobenzhydrol |
| Smältpunkt | 91°C to 93°C |
| Kvantitet | 5 g |
| Beilstein | 1878717 |
| Löslighetsinformation | Insoluble in water. |
| Formel vikt | 253.13 |
| Procent renhet | 98% |