Läs mer
4,4'-Dichlorobenzhydrol, TRC
Produktkod. 30369298
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30369298 5 g
1 options
Produktkod. 30369298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4365985G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material DBH
CAS 90-97-1
Molekylformel C13 H10 Cl2 O
Kvantitet 5 g
Synonym Benzenemethanol, 4-chloro-α-(4-chlorophenyl)- (9CI, ACI), 4-Chloro-α-(4-chlorophenyl)benzenemethanol (ACI), Benzhydrol, 4,4'-dichloro- (6CI, 7CI, 8CI), 4,4'-Dichlorobenzhydrol, 4,4'-Dichlorobenzhydryl alcohol, 4,4'-Dichlorodiphenylcarbinol, Bis(4-chlorophenyl)methanol, Bis(p-chlorophenyl)methanol, DBH, DBH (degradation product), NSC 121779, NSC 5250, p,p'-Dichlorobenzhydrol
InChI formel InChI=1S/C13H10Cl2O/c14-11-5-1-9(2-6-11)13(16)10-3-7-12(15)8-4-10/h1-8,13,16H
LEDER OC(c1ccc(Cl)cc1)c2ccc(Cl)cc2
IUPAC-namn bis(4-chlorophenyl)methanol
Molekylvikt (g/mol) 253.12
Formel vikt 252.0109