Läs mer
4,4'-Dichlorodiphenyltrichloroethane, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4,4'-DDT |
| CAS | 50-29-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C14 H9 Cl5 |
| Kvantitet | 250 mg |
| Synonym | Ethane, 1,1,1-trichloro-2,2-bis(p-chlorophenyl)- (7CI,8CI), 1,1'-(2,2,2-Trichloroethylidene)bis[4-chlorobenzene], 1,1,1-Trichloro-2,2-bis(p-chlorophenyl)ethane, 1,1,1-Trichloro-2,2-di(p-chlorophenyl)ethane, 1,1,1-Trichloro-2-2,2-bis(p-chlorophenyl)ethane, 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethane, 1,1-Bis(p-chlorophenyl)-2,2,2-trichloroethane, 2,2,2-Trichloro-1,1-bis(4-chlorophenyl)ethane, 2,2-Bis(4-chlorophenyl)-1,1,1-trichloroethane, 2,2-Bis(p-chlorophenyl)-1,1,1-trichloroethane, 4,4'-DDT, 4,4'-Dichlorodiphenyltrichloroethane, Aavero-extra, Agritan, Arkotine, Azotox M 33, Benzochloryl, Bosan supra, Bovidermol, Chlorophenothane, Chlorphenothan, Chlorphenotoxum, Citox, Clofenotan, Clofenotane, DDT, Deoval, Detox, Detox (pesticide), Detoxan, Dibovin, Dichlorodiphenyltrichloroethane, Dicophane, Dicophaner, Dodat, Dykol, ENT-1506, Estonate, Ethane, 1,1,1-trichloro-2,2-bis(4-chlorophenyl)-, FWJ, Gesafid, Gesapon, Gesarex, Gesarol, Guesapon, Hildit, Ivoran, Mutoxan, NSC 8939, Neocid, Neocidol, Neocidol (solid), PEB1, Parachlorocidum, Pentachlorin, Penticidum, Tafidex, Trichlorobis(4'-chlorophenyl)ethane, Zerdane, o,p'-1,1,1-Trichloro-2-2,2-bis(p-chlorophenyl)ethane, p,p'-DDT, p,p'-Dichlorodiphenyltrichloroethane, p,p'-Dichlorodiphenyltrichloromethylmethane, α,α-Bis(p-chlorophenyl)-β,β,β-trichlorethane |
| InChI formel | InChI=1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H |
| LEDER | Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl |
| IUPAC-namn | 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene |
| Molekylvikt (g/mol) | 354.49 |
| Visa mer |