Läs mer
4,4'-Dicyanobenzophenone, TRC
Produktkod. 17950795
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17950795 250 mg
17933005 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17950795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD436880250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32446-66-5
Molekylformel C15 H8 N2 O
Molekylvikt (g/mol) 232.24
Synonym Benzonitrile, 4,4'-carbonylbis-, Benzonitrile, 4,4'-carbonyldi- (8CI), 4,4'-Carbonylbis[benzonitrile], 4,4'-Dicyanobenzophenone, 4-[(4-Cyanophenyl)carbonyl]benzonitrile, Bis(4-cyanophenyl) ketone, Bis(p-cyanophenyl) ketone, p,p'-Dicyanobenzophenone
IUPAC-namn 4-(4-cyanobenzoyl)benzonitrile
LEDER O=C(c1ccc(cc1)C#N)c2ccc(cc2)C#N
Specifikationer
Kemiskt namn eller material 4,4'-Carbonyldibenzonitrile
Kvantitet 250 mg
InChI formel InChI=1S/C15H8N2O/c16-9-11-1-5-13(6-2-11)15(18)14-7-3-12(10-17)4-8-14/h1-8H
Formel vikt 232.0637