Läs mer
4,4-Diethoxy-N,N-diethyl-1-butanamine, TRC
Produktkod. 30349220
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349220 25 mg
30350483 100 mg
30351518 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD16897025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97028-91-6
Molekylformel C12H27NO2
Molekylvikt (g/mol) 217.35
Synonym 4-(Diethylamino)butanal Diethyl Acetal
IUPAC-namn 4,4-diethoxy-N,N-diethylbutan-1-amine
LEDER CCOC(CCCN(CC)CC)OCC
Specifikationer
Kemiskt namn eller material 4,4-Diethoxy-N,N-diethyl-1-butanamine
Kvantitet 25 mg
InChI formel InChI=1S/C12H27NO2/c1-5-13(6-2)11-9-10-12(14-7-3)15-8-4/h12H,5-11H2,1-4H3
Formel vikt 217.2042