missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diethylbiphenyl, TRC

Product Code. 30373451
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30373451 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373451 Supplier Toronto Research Chemicals Supplier No. TRCD444878250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4,4'-Diethylbiphenyl
CAS 13049-40-6
Molekylformel C16H18
Kvantitet 250 mg
InChI formel InChI=1S/C16H18/c1-3-13-5-9-15(10-6-13)16-11-7-14(4-2)8-12-16/h5-12H,3-4H2,1-2H3
LEDER CCc1ccc(cc1)c2ccc(CC)cc2
IUPAC-namn 1-ethyl-4-(4-ethylphenyl)benzene
Molekylvikt (g/mol) 210.31
Formel vikt 210.1409
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.