Learn More
4,4'-Difluorobenzhydryl Bromide, TRC
Product Code. 30004626
Change view
Click to view available options
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30004626 1 g
30003747 500 mg
30006710 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004626 Supplier Toronto Research Chemicals Supplier No. TRCB5191051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 345-90-4
Molekylformel C13H9BrF2
Molekylvikt (g/mol) 283.11
Synonym Bis(4-fluorophenyl)bromomethane, Bis(4-fluorophenyl)methyl Bromide, Bromobis(p-fluorophenyl)methane, alpha-Bromobis(p-fluorophenyl)methane, 1,1'-(Bromomethylene)bis[4-fluorobenzene], 1-[Bromo-(4-fluorophenyl)methyl]-4-fluorobenzene, 4,4'-(Bromomethylene)bis(fluorobenzene);
IUPAC-namn 1-[bromo-(4-fluorophenyl)methyl]-4-fluorobenzene
LEDER Fc1ccc(cc1)C(Br)c2ccc(F)cc2
Specifications
Kemiskt namn eller material 4,4'-Difluorobenzhydryl Bromide
Kvantitet 1 g
InChI formel InChI=1S/C13H9BrF2/c14-13(9-1-5-11(15)6-2-9)10-3-7-12(16)8-4-10/h1-8,13H
Formel vikt 281.9856