Läs mer
4,4'-Dimethoxy Diphenyl Sulfide, TRC
Produktkod. 30374434
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374434 500 mg
30377184 50 mg
30368858 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30374434 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD460998500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3393-77-9
Molekylformel C14H14O2S
Molekylvikt (g/mol) 246.32
LEDER COc1ccc(Sc2ccc(OC)cc2)cc1
Specifikationer
Kemiskt namn eller material 4,4'-Dimethoxy Diphenyl Sulfide
Kvantitet 500 mg
InChI formel InChI=1S/C14H14O2S/c1-15-11-3-7-13(8-4-11)17-14-9-5-12(16-2)6-10-14/h3-10H,1-2H3
Formel vikt 246.0715