Learn More
4,4'-Dimethoxytrityl chloride, 98%, BLD Pharm™
Product Code. 30985558
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30985558 5 g
30980221 25 g
30982308 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30985558 Supplier BLD Pharm Supplier No. BD13515g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 40615-36-9
Molecular Formula C21H19ClO2
MDL Number MFCD00008409
Quantity 5 g
SMILES C3=C(C(C1=CC=C(OC)C=C1)(C2=CC=CC=C2)Cl)C=CC(=C3)OC
Molecular Weight (g/mol) 338.83
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethoxytrityl chloride