Läs mer
4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione, TRC
Produktkod. 30387701
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30387701 10 mg
30378543 50 mg
30385688 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30387701 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45797810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5488-36-8
Molekylformel C11H11NO2
Molekylvikt (g/mol) 189.21
LEDER CC1(C)C(=O)NC(=O)c2ccccc12
Specifikationer
Kemiskt namn eller material 4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione
Kvantitet 10 mg
InChI formel InChI=1S/C11H11NO2/c1-11(2)8-6-4-3-5-7(8)9(13)12-10(11)14/h3-6H,1-2H3,(H,12,13,14)
Formel vikt 189.079