Läs mer
4,4-Dimethyl-1,3-dioxolan-2-one, TRC
Produktkod. 30373975
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30373975 5 g
1 options
Produktkod. 30373975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4717905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4-Dimethyl-1,3-dioxolan-2-one
CAS 4437-69-8
Molekylformel C5H8O3
Kvantitet 5 g
Synonym Carbonic acid 1,1-Dimethylethylene Ester, Carbonic Acid Cyclic 1,1-Dimethylethylene Ester, 2-Methyl-1,2-propanediol Cyclic Carbonate, 1,1-Dimethylethylene Carbonate, Isobutylene Carbonate
InChI formel InChI=1S/C5H8O3/c1-5(2)3-7-4(6)8-5/h3H2,1-2H3
LEDER CC1(C)COC(=O)O1
IUPAC-namn 4,4-dimethyl-1,3-dioxolan-2-one
Molekylvikt (g/mol) 116.12
Formel vikt 116.0473