missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethyl-2-pentyne, 97+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Description
4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Density | 0.718 |
Boiling Point | 82°C |
Flash Point | −10°C (14°F) |
Odor | Petrol-like |
Linear Formula | (CH3)3CC≡CCH3 |
Refractive Index | 1.4085 |
Quantity | 1 g |
UN Number | UN3295 |
Beilstein | 1733540 |
Solubility Information | Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'