Läs mer
4-[4-(Dimethylamino)-1-oxobutyl]-3-(hydroxymethyl)benzonitrile, TRC
Produktkod. 30379106
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379106 1 mg
30373264 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379106 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4617451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1433278-33-1
Molekylformel C14H18N2O2
Molekylvikt (g/mol) 246.3
Synonym Benzonitrile, 4-[4-(dimethylamino)-1-oxobutyl]-3-(hydroxymethyl)-, 4-[4-(Dimethylamino)butanoyl]-3-(hydroxymethyl)benzonitrile
LEDER CN(C)CCCC(=O)c1ccc(cc1CO)C#N
Specifikationer
Kemiskt namn eller material 4-[4-(Dimethylamino)-1-oxobutyl]-3-(hydroxymethyl)benzonitrile
Kvantitet 1 mg
InChI formel InChI=1S/C14H18N2O2/c1-16(2)7-3-4-14(18)13-6-5-11(9-15)8-12(13)10-17/h5-6,8,17H,3-4,7,10H2,1-2H3
Formel vikt 246.1368