Läs mer
4,4-Dimethylpentan-2-ol, TRC
Produktkod. 30382271
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30382271 50 mg
30381975 100 mg
30372643 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30382271 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45594550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6144-93-0
Molekylformel C7H16O
Molekylvikt (g/mol) 116.2
IUPAC-namn 4,4-dimethylpentan-2-ol
LEDER CC(O)CC(C)(C)C
Specifikationer
Kemiskt namn eller material 4,4-Dimethylpentan-2-ol
Kvantitet 50 mg
InChI formel InChI=1S/C7H16O/c1-6(8)5-7(2,3)4/h6,8H,5H2,1-4H3
Formel vikt 116.1201