Läs mer
4,4'-Dithiobisbenzoic Acid Dimethyl Ester, TRC
Produktkod. 30371596
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30371596 1 g
1 options
Produktkod. 30371596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4942101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4'-Dithiobisbenzoic Acid Dimethyl Ester
CAS 35190-68-2
Molekylformel C16H14O4S2
Kvantitet 1 g
Synonym 4,4'-Dithiodibenzoic Acid Dimethyl Ester, Bis(4-carbomethoxyphenyl) Disulfide
InChI formel InChI=1S/C16H14O4S2/c1-19-15(17)11-3-7-13(8-4-11)21-22-14-9-5-12(6-10-14)16(18)20-2/h3-10H,1-2H3
LEDER COC(=O)c1ccc(SSc2ccc(cc2)C(=O)OC)cc1
Molekylvikt (g/mol) 334.41
Formel vikt 334.0334