Learn More
4-(4-Fluoro-2-nitrophenyl)morpholine, 97%, Thermo Scientific™
Catalog Number 15489838
Click to view available options
Quantity:
1 g
5 g
25 g
238418-75-2
C10H11FN2O3
226.21
MFCD00798369
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
2737431
4-(4-fluoro-2-nitrophenyl)morpholine
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCOCC1
238418-75-2
C10H11FN2O3
226.21
MFCD00798369
XEVFYVWDTJTEHQ-UHFFFAOYSA-N
4-4-fluoro-2-nitrophenyl morpholine, acmc-1cbj8, 5-fluoro-2-morpholinonitrobenzene, 4-4-fluoro-2-nitro-phenyl-morpholine, morpholine,4-4-fluoro-2-nitrophenyl
2737431
4-(4-fluoro-2-nitrophenyl)morpholine
[O-][N+](=O)C1=C(C=CC(F)=C1)N1CCOCC1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 226.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Fluoro-2-nitrophenyl)morpholine