missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-4812999
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5731-10-2
LXWNTLBMNCXRQN-UHFFFAOYSA-M
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
2782689
This item is not returnable. View return policy
5731-10-2
LXWNTLBMNCXRQN-UHFFFAOYSA-M
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
2782689
Melting Point 241.0°C to 243.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C13H9FO2
Quantity 1 g
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
IUPAC Name 4-(4-fluorophenyl)benzoic acid
Molecular Weight (g/mol) 216.21
Formula Weight 216.21
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenyl)benzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4-Fluorophenyl)benzoic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'