Learn More
4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 166.0°C to 169.5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 5 g |
Synonym | 4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid |
Formula Weight | 268.27 |
Percent Purity | 97% |
Chemical Name or Material | 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid |
Safety and Handling
Product Identifier
- 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.