missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%, ACROS Organics™

Brand:  Acros Organics 454520250

Additional Details : CAS Number : 197304-21-5 Weight : 0.02500kg


Quantity 25g

 View more versions of this product

Product Code. 15204368

  • 10407.82 SEK / 25g


Estimated Shipment
06-11-2020
Add to basket


Specifications

Chemical Identifiers

197304-21-5
268.27
4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid
4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid
C13H16O6
SEJSVFHBVLKHLB-UHFFFAOYSA-N
2758419
COC1=CC(=CC(=C1C=O)OC)OCCCC(=O)O

Specifications

4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid
166.0°C to 169.5°C
197304-21-5
C13H16O6
SEJSVFHBVLKHLB-UHFFFAOYSA-N
4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid
2758419
97%
Authentic
25g
96% min. (HPLC)
4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid
COC1=CC(=CC(=C1C=O)OC)OCCCC(=O)O
268.27
268.27
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

Documents