Learn More
4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%, Thermo Scientific™
625.99 SEK - 10299.63 SEK
Chemical Identifiers
CAS | 197304-21-5 |
---|---|
Molecular Formula | C13H16O6 |
Molecular Weight (g/mol) | 268.27 |
MDL Number | MFCD01317805 |
InChI Key | SEJSVFHBVLKHLB-UHFFFAOYSA-N |
Synonym | 4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid |
PubChem CID | 2758419 |
IUPAC Name | 4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid |
SMILES | COC1=CC(OCCCC(O)=O)=CC(OC)=C1C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15204368
|
Thermo Scientific Acros
454520250 |
25g |
10299.63 SEK
25g |
Estimated Shipment: 01-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15294358
|
Thermo Scientific Acros
454520050 |
5g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
15258336
|
Thermo Scientific Acros
454520010 |
1g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Chemical Identifiers
197304-21-5 | |
268.27 | |
SEJSVFHBVLKHLB-UHFFFAOYSA-N | |
2758419 | |
COC1=CC(OCCCC(O)=O)=CC(OC)=C1C=O |
C13H16O6 | |
MFCD01317805 | |
4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid | |
4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid |
Specifications
4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid | |
166.0°C to 169.5°C | |
Authentic | |
MFCD01317805 | |
25g | |
COC1=CC(OCCCC(O)=O)=CC(OC)=C1C=O | |
268.27 | |
268.27 |
197304-21-5 | |
96% min. (HPLC) | |
C13H16O6 | |
4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid | |
SEJSVFHBVLKHLB-UHFFFAOYSA-N | |
4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid | |
2758419 | |
97% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only