Läs mer
4,4'-Iminodiphenol, TRC
Produktkod. 30136424
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136424 100 mg
30007033 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI409010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1752-24-5
Molekylformel C12H11NO2
Molekylvikt (g/mol) 201.22
Synonym 4,4'-Iminobisphenol, Phenol, 4,4'-iminodi- (6CI,7CI,8CI), 4,4'-Iminobis[phenol], 4,4'-Dihydroxydiphenylamine, 4,4'-Iminobisphenol, 4,4'-Iminodiphenol, 4-(4-Hydroxyanilino)phenol, Bis(4-hydroxyphenyl)amine, Bis(p-hydroxyphenyl)amine, Indophenol, reduced, Leucoindophenol, N-(4'-Hydroxyphenyl)-p-aminophenol, 4,4'-Azanediyldiphenol, Paracetamol Imp. M (EP)
IUPAC-namn 4-(4-hydroxyanilino)phenol
LEDER Oc1ccc(Nc2ccc(O)cc2)cc1
Specifikationer
Kemiskt namn eller material 4,4'-Iminodiphenol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H11NO2/c14-11-5-1-9(2-6-11)13-10-3-7-12(15)8-4-10/h1-8,13-15H
Formel vikt 201.079
Procent renhet >95%