missing translation for 'onlineSavingsMsg'
Learn More

4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034233
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1448637-05-5
C22H23BO4
362.23
MFCD16294527
SGGJQXOMPWFXNR-UHFFFAOYSA-N
methyl 4-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-benzoic acid methyl ester, amtb207, 4-4-methoxycarbonyl phenyl ethynyl benzeneboronic acid pinacol ester, methyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl benzoic acid methyl ester
71306515
methyl 4-[2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl]benzoate
COC(=O)C1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
1448637-05-5
C22H23BO4
362.23
MFCD16294527
SGGJQXOMPWFXNR-UHFFFAOYSA-N
methyl 4-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl-benzoic acid methyl ester, amtb207, 4-4-methoxycarbonyl phenyl ethynyl benzeneboronic acid pinacol ester, methyl 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethynyl benzoate, 4-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylethynyl benzoic acid methyl ester
71306515
methyl 4-[2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethynyl]benzoate
COC(=O)C1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 362.23
Percent Purity 95%
Chemical Name or Material 4-([4-(Methoxycarbonyl)phenyl]ethynyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'