Läs mer
4,4'-Oxybis(butan-1-ol), TRC
Produktkod. 30410936
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410936 50 mg
30410302 10 mg
30410044 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410936 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO86854350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3403-82-5
Molekylformel C8H18O3
Molekylvikt (g/mol) 162.22
Synonym Dibutylene Glycol
IUPAC-namn 4-(4-hydroxybutoxy)butan-1-ol
LEDER OCCCCOCCCCO
Specifikationer
Kemiskt namn eller material 4,4'-Oxybis(butan-1-ol)
Kvantitet 50 mg
InChI formel InChI=1S/C8H18O3/c9-5-1-3-7-11-8-4-2-6-10/h9-10H,1-8H2
Formel vikt 162.1256