missing translation for 'onlineSavingsMsg'
Läs mer

4-(4-Phenylbutoxy)benzoic Acid, TRC

Produktkod. 30240877
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240877 25 mg
30240705 50 mg
30240823 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30240877 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP39925025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 30131-16-9
Molekylformel C17H18O3
Molekylvikt (g/mol) 270.32
IUPAC-namn 4-(4-phenylbutoxy)benzoic acid
LEDER OC(=O)c1ccc(OCCCCc2ccccc2)cc1

Specifications

Kemiskt namn eller material 4-(4-Phenylbutoxy)benzoic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C17H18O3/c18-17(19)15-9-11-16(12-10-15)20-13-5-4-8-14-6-2-1-3-7-14/h1-3,6-7,9-12H,4-5,8,13H2,(H,18,19)
Formel vikt 270.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.