Läs mer
4,4'-(Pyridin-2-ylmethylene)diphenol, Mikromol™
Produktkod. 17928954
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17928954 25 mg
17918384 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17928954 Leverantör Mikromol Leverantörsnummer MM0031.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4,4'-(Pyridin-2-ylmethylene)diphenol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4,4'-(Pyridin-2-ylmethylene)diphenol
CAS 603-41-8
Analyt- eller komponentnamn 4,4'-(Pyridin-2-ylmethylene)diphenol
Frakt skick Room Temperature
Molekylformel C18 H15 N O2
Certifikat och efterlevnad ISO 17025
Synonym Bisacodyl Imp. A (EP), Sodium Picosulfate Imp. B (EP), Bisacodyl USP Related Compound A, Bisacodyl USP RC A, 4,4'-[(Pyridin-2-yl)methylene]diphenol, 4,4'-(Pyridin-2-ylmethylene)diphenol, Bisacodyl Related Compound A, Bisacodyl Impurity A, Sodium Picosulfate Impurity B, Sodium Picosulfate Impurity B
InChI formel InChI=1S/C18H15NO2/c20-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(21)11-7-14/h1-12,18,20-21H
LEDER Oc1ccc(cc1)C(c2ccc(O)cc2)c3ccccn3
IUPAC-namn 4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenol
Molekylvikt (g/mol) 277.32
Kvantitet 25 mg
Formel vikt 277.1103 g/mol