missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Pyridyl)-2-(3-pyridylmethyl)thiazole, 97%, Thermo Scientific™

Catalog Number p-7038963
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1421263-26-4
C14H11N3S
253.323
MFCD22682928
KUCSSSQZPFTBHS-UHFFFAOYSA-N
4-4-pyridyl-2-3-pyridylmethyl thiazole, 4-2-pyridin-3-ylmethyl-1,3-thiazol-4-yl pyridine, 2-pyridin-3-ylmethyl-4-pyridin-4-yl thiazole
71741367
4-pyridin-4-yl-2-(pyridin-3-ylmethyl)-1,3-thiazole
C1=CC(=CN=C1)CC2=NC(=CS2)C3=CC=NC=C3
This item is not returnable. View return policy
1421263-26-4
C14H11N3S
253.323
MFCD22682928
KUCSSSQZPFTBHS-UHFFFAOYSA-N
4-4-pyridyl-2-3-pyridylmethyl thiazole, 4-2-pyridin-3-ylmethyl-1,3-thiazol-4-yl pyridine, 2-pyridin-3-ylmethyl-4-pyridin-4-yl thiazole
71741367
4-pyridin-4-yl-2-(pyridin-3-ylmethyl)-1,3-thiazole
C1=CC(=CN=C1)CC2=NC(=CS2)C3=CC=NC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 253.32
Percent Purity 97%
Chemical Name or Material 4-(4-Pyridyl)-2-(3-pyridylmethyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'