Learn More
4,4′-Thiobis(2-tert-butyl-5-methylphenol), Sigma-Aldrich™
Specifications
Specifications
| CAS | 96-69-5 |
| Melting Point | 158°C to 162°C |
| Molecular Formula | (2-(CH3)-4-(OH)-5-((CH3)3C)C6H2)2S |
| MDL Number | MFCD00026287 |
| Quantity | 100 g |
| Synonym | 4,4′-Thiobis(2-tert-butyl-5-methylphenol), Bis(3-tert-butyl-4-hydroxy-6-methylphenyl)sulfide, 4,4′-Thiobis(2-tert-butyl-5-methylphenol) |
| Molecular Weight (g/mol) | 358.54 |
| Grade | Research |