missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 1160058-86-5 |
Assay Percent Range | 97% |
Molecular Formula | C11H6ClF3N2O3S |
Quantity | 1 g |
Synonym | 4-4-trifluoromethyl pyrimidin-2-yl oxy benzenesulfonyl chloride, 4-4-trifluoromethyl pyrimidin-2-yl oxy benzene-1-sulfonyl chloride, 4-4-trifluoromethyl-2-pyrimidinyloxy benzenesulfonyl chloride, 4-4-trifluoromethylpyrimidin-2-oxy benzenesulfonyl chloride |
InChI Key | SMDWXDCIMUGFNP-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1OC2=NC=CC(=N2)C(F)(F)F)S(=O)(=O)Cl |
IUPAC Name | 4-[4-(trifluoromethyl)pyrimidin-2-yl]oxybenzenesulfonyl chloride |
Molecular Weight (g/mol) | 338.685 |
PubChem CID | 45594321 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'